Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032565.hg.1 |
TC02002037.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032565.hg.1 |
ENST00000263854.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032565.hg.1 |
ENST00000295802.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032565.hg.1 |
ENST00000457495.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02032565.hg.1 |
NM_017750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200196844.hg.1 |
ENST00000295802.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196844.hg.1 |
ENST00000409984.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196844.hg.1 |
ENST00000482694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196844.hg.1 |
NM_017750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200196844.hg.1 |
ENST00000295802.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196844.hg.1 |
ENST00000409984.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196844.hg.1 |
ENST00000482694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200196844.hg.1 |
NM_017750.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200196844.hg.1 |
TC0200013261.hg.1 |
Clariom D TC |