Assay Details
Gene Symbol: SSC5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144950.1 | NP_001138422.1 | 13 - 14 | 3029 | 64 | |
GenBank mRNA | EU699476.1 | - | 13 - 14 | 2965 | 64 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000900.hg.1
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
receptor-mediated endocytosis
multicellular organism development
detection of bacterial lipoprotein
innate immune response
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
negative regulation of interleukin-8 secretion
fibronectin binding
scavenger receptor activity
protein binding
laminin binding
extracellular matrix binding
receptor-mediated endocytosis
multicellular organism development
detection of bacterial lipoprotein
innate immune response
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
negative regulation of interleukin-8 secretion
fibronectin binding
scavenger receptor activity
protein binding
laminin binding
extracellular matrix binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19012497.hg.1 | ENST00000389623.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19012497.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19012497.hg.1 | TC19000900.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000589020.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000589020.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000594321.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000594321.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000594321.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000594321.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000594321.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900169891.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900169891.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900169891.hg.1 | TC1900008912.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900169891.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900169891.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000389623.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000587166.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000588254.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000589020.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000589020.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000594321.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000594321.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000594321.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000594321.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000594321.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | ENST00000615233.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001144950.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008912.hg.1 | NM_001195267.1 | Refseq |
