Assay Details
| Gene Symbol: MYO5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018728.3 | NP_061198.2 | 28 - 29 | 3608 | 86 | |
| XM_011521781.2 | XP_011520083.1 | 28 - 29 | 3524 | 86 | ||
| XM_017022408.1 | XP_016877897.1 | 27 - 28 | 3473 | 86 | ||
| XM_017022409.1 | XP_016877898.1 | 23 - 24 | 2915 | 86 | ||
| XM_017022410.1 | XP_016877899.1 | 27 - 28 | 3458 | 86 | ||
| GenBank mRNA | AF161352.1 | - | 7 - 8 | 607 | 86 | |
| AF272390.1 | - | 28 - 29 | 3465 | 86 | ||
| BC144611.1 | - | 26 - 27 | 3163 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001454.hg.1
Set Membership: |
|

MYO5C
Genomic Map