Assay Details
| Gene Symbol: MYO5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018728.3 | NP_061198.2 | 31 - 32 | 3948 | 80 | |
| XM_011521781.2 | XP_011520083.1 | 31 - 32 | 3864 | 80 | ||
| XM_017022408.1 | XP_016877897.1 | 30 - 31 | 3813 | 80 | ||
| XM_017022409.1 | XP_016877898.1 | 26 - 27 | 3255 | 80 | ||
| XM_017022410.1 | XP_016877899.1 | 30 - 31 | 3798 | 80 | ||
| GenBank mRNA | AF272390.1 | - | 31 - 32 | 3805 | 80 | |
| BC144611.1 | - | 29 - 30 | 3503 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001454.hg.1
Set Membership: |
|

MYO5C
Genomic Map