Assay Details
| Gene Symbol: LARP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286679.1 | NP_001273608.1 | 2 - 3 | 330 | 74 | |
| NM_018357.3 | NP_060827.2 | 2 - 3 | 595 | 74 | ||
| GenBank mRNA | AF443828.1 | - | 2 - 3 | 488 | 74 | |
| AF443829.1 | - | 2 - 3 | 488 | 74 | ||
| AK002058.1 | - | 2 - 3 | 489 | 74 | ||
| AK130092.1 | - | 3 - 4 | 389 | 74 | ||
| AL833876.1 | - | 2 - 3 | 449 | 74 | ||
| BC006082.1 | - | 2 - 3 | 442 | 2961587 | 74 | |
| BC009446.1 | - | 2 - 3 | 442 | 3529516 | 74 | |
| BC014018.2 | - | 2 - 3 | 446 | 4127153 | 74 | |
| BI913258.1 | - | 2 - 3 | 503 | 74 | ||
| DA715263.1 | - | 2 - 3 | 330 | 74 | ||
| DN992782.1 | - | 2 - 3 | 600 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001286679.1 transcripts.
Set Membership: |
|

LARP6
Genomic Map