Assay Details
| Gene Symbol: HUNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014586.1 | NP_055401.1 | 9 - 10 | 1663 | 114 | |
| GenBank mRNA | AJ271722.1 | - | 9 - 10 | 1663 | 114 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000113.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000430354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000430354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000430354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000430354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000439107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000439107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000439107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000439107.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000465574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000465574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000465574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000465574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | ENST00000465574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000716.hg.1 | ENST00000270112.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000716.hg.1 | ENST00000465574.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000716.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21000716.hg.1 | TC21000113.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2100146430.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146430.hg.1 | ENST00000439107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146430.hg.1 | ENST00000465574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146430.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100146430.hg.1 | TC2100006909.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100146430.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146430.hg.1 | ENST00000439107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146430.hg.1 | ENST00000465574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100146430.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000270112.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000430354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000430354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000430354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000430354.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000439107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000439107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000439107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000439107.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000465574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000465574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000465574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000465574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | ENST00000465574.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | NM_014586.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006909.hg.1 | NM_014586.1 | Refseq |

HUNK
Genomic Map