Assay Details
| Gene Symbol: EIF5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020390.5 | NP_065123.1 | 1 - 2 | 97 | 109 | |
| GenBank mRNA | AK311962.1 | - | 1 - 2 | 58 | 109 | |
| AY205258.1 | - | 1 - 2 | 97 | 109 | ||
| AY205259.1 | - | 1 - 2 | 97 | 109 | ||
| AY205260.1 | - | 1 - 2 | 97 | 109 | ||
| AY205261.1 | - | 1 - 2 | 97 | 109 | ||
| BC036072.1 | - | 1 - 2 | 68 | 5288175 | 109 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001995.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK311962.1,AY205258.1,AY205259.1,AY205260.1,AY205261.1,BC036072.1,NM_020390.5 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
translational frameshifting
spermatogenesis
positive regulation of cell proliferation
peptidyl-lysine modification to peptidyl-hypusine
polyamine homeostasis
protein transport
positive regulation of translational elongation
positive regulation of translational termination
mRNA transport
translation elongation factor activity
protein binding
ribosome binding
translational frameshifting
spermatogenesis
positive regulation of cell proliferation
peptidyl-lysine modification to peptidyl-hypusine
polyamine homeostasis
protein transport
positive regulation of translational elongation
positive regulation of translational termination
mRNA transport
translation elongation factor activity
protein binding
ribosome binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500009948.hg.1 | ENST00000564027.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035669.hg.1 | TC03001995.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035669.hg.1 | ENST00000295822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035669.hg.1 | ENST00000474096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035669.hg.1 | ENST00000487522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035669.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000474417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000474417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000474417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000474417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000474417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035668.hg.1 | TC03001995.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035668.hg.1 | ENST00000295822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035668.hg.1 | ENST00000474096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035668.hg.1 | ENST00000487522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035668.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035670.hg.1 | TC03001995.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035670.hg.1 | ENST00000295822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035670.hg.1 | ENST00000460117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035670.hg.1 | ENST00000474096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035670.hg.1 | ENST00000487522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035670.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035673.hg.1 | TC03001995.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035673.hg.1 | ENST00000295822.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035673.hg.1 | ENST00000460117.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035673.hg.1 | ENST00000474096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035673.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000474417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000474417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000474417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000474417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000474417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013104.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198005.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198005.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198005.hg.1 | ENST00000474417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198005.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198005.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198005.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198005.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198005.hg.1 | ENST00000474417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198005.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198005.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198005.hg.1 | TC0300013104.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300198011.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198011.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198011.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198011.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198011.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198011.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198011.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198011.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198011.hg.1 | TC0300013104.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300198007.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198007.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198007.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198007.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198007.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198007.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198007.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198007.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198007.hg.1 | TC0300013104.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500009948.hg.1 | ENST00000564027.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198008.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198008.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198008.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198008.hg.1 | ENST00000474417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198008.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198008.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198008.hg.1 | ENST00000295822.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198008.hg.1 | ENST00000460117.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198008.hg.1 | ENST00000474096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198008.hg.1 | ENST00000474417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198008.hg.1 | ENST00000487522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198008.hg.1 | NM_020390.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198008.hg.1 | TC0300013104.hg.1 | Clariom D TC |

EIF5A2
Genomic Map