Assay Details
| Gene Symbol: FAM186B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032130.2 | NP_115506.1 | 3 - 4 | 683 | 67 | |
| NR_027450.1 | 3 - 4 | 840 | 67 | |||
| XM_006719625.2 | XP_006719688.1 | 3 - 4 | 672 | 67 | ||
| XM_006719626.2 | XP_006719689.1 | 3 - 4 | 672 | 67 | ||
| XM_006719627.3 | XP_006719690.1 | 3 - 4 | 672 | 67 | ||
| XM_011538796.2 | XP_011537098.1 | 3 - 4 | 672 | 67 | ||
| XM_011538797.2 | XP_011537099.1 | 3 - 4 | 672 | 67 | ||
| XM_017020008.1 | XP_016875497.1 | 3 - 4 | 923 | 67 | ||
| GenBank mRNA | AK093242.1 | - | 3 - 4 | 672 | 67 | |
| AK302702.1 | - | 3 - 4 | 840 | 67 | ||
| AL136748.1 | - | 3 - 4 | 604 | 67 | ||
| BC035621.1 | - | 3 - 4 | 641 | 5171752 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001480.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000527253.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000527253.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000547764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000547764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000547764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000547764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000552301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000552301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000552301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000552301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019432.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019432.hg.1 | ENST00000544141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019432.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019432.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019432.hg.1 | TC12001480.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019433.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019433.hg.1 | ENST00000544141.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019433.hg.1 | ENST00000551047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019433.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019433.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12019433.hg.1 | TC12001480.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200180900.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180900.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180900.hg.1 | ENST00000551047.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180900.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180900.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180900.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180900.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180900.hg.1 | ENST00000551047.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180900.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180900.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180900.hg.1 | TC1200010635.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000527253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000527253.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000547764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000547764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000547764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000547764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000552301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000552301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000552301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000552301.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180898.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180898.hg.1 | ENST00000532262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180898.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180898.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180898.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180898.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180898.hg.1 | ENST00000532262.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180898.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180898.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180898.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180898.hg.1 | TC1200010635.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200180899.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180899.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180899.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180899.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180899.hg.1 | ENST00000257894.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180899.hg.1 | ENST00000533372.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200180899.hg.1 | NM_032130.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180899.hg.1 | NR_027450.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200180899.hg.1 | TC1200010635.hg.1 | Clariom D TC |

FAM186B
Genomic Map