Assay Details
| Gene Symbol: STX1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052874.4 | NP_443106.1 | 2 - 3 | 348 | 72 | |
| XM_017022893.1 | XP_016878382.1 | 2 - 3 | 103 | 72 | ||
| GenBank mRNA | AY028792.1 | - | 2 - 3 | 164 | 72 | |
| AY995211.1 | - | 2 - 3 | 106 | 72 | ||
| BC062298.1 | - | 2 - 3 | 160 | 6375788 | 72 | |
| BX102270.1 | - | 4 - 5 | 72 | |||
| DN990202.1 | - | 2 - 3 | 342 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001047.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of neurotransmitter secretion
intracellular protein transport
regulation of gene expression
regulation of synaptic vesicle priming
synaptic vesicle docking
synaptic vesicle fusion to presynaptic active zone membrane
vesicle docking
spontaneous neurotransmitter secretion
positive regulation of excitatory postsynaptic potential
SNARE binding
SNAP receptor activity
protein kinase binding
protein domain specific binding
positive regulation of neurotransmitter secretion
intracellular protein transport
regulation of gene expression
regulation of synaptic vesicle priming
synaptic vesicle docking
synaptic vesicle fusion to presynaptic active zone membrane
vesicle docking
spontaneous neurotransmitter secretion
positive regulation of excitatory postsynaptic potential
SNARE binding
SNAP receptor activity
protein kinase binding
protein domain specific binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16014903.hg.1 | ENST00000215095.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014903.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014903.hg.1 | TC16001047.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000566211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000566211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000566211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014905.hg.1 | ENST00000215095.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014905.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014905.hg.1 | TC16001047.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014907.hg.1 | ENST00000215095.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014907.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014907.hg.1 | TC16001047.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014904.hg.1 | ENST00000215095.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014904.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014904.hg.1 | TC16001047.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014902.hg.1 | ENST00000215095.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014902.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014902.hg.1 | TC16001047.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014906.hg.1 | ENST00000215095.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014906.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014906.hg.1 | TC16001047.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600177311.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177311.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177311.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177311.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177311.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177311.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177311.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177311.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177311.hg.1 | TC1600010000.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600177313.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177313.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177313.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177313.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177313.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177313.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177313.hg.1 | TC1600010000.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600177314.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177314.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177314.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177314.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177314.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177314.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177314.hg.1 | TC1600010000.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600177315.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177315.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177315.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177315.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177315.hg.1 | TC1600010000.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000566211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000566211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000566211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | ENST00000569638.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010000.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177310.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177310.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177310.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177310.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177310.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177310.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177310.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177310.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177310.hg.1 | TC1600010000.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600177312.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177312.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177312.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177312.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177312.hg.1 | ENST00000215095.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177312.hg.1 | ENST00000561836.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177312.hg.1 | ENST00000565419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600177312.hg.1 | NM_052874.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600177312.hg.1 | TC1600010000.hg.1 | Clariom D TC |

STX1B
Genomic Map