Assay Details
| Gene Symbol: ANXA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003954.2 | NP_001003954.1 | 3 - 4 | 304 | 92 | |
| NM_004306.3 | NP_004297.2 | 2 - 3 | 181 | 92 | ||
| GenBank mRNA | AJ306450.1 | - | 3 - 4 | 280 | 92 | |
| AK223374.1 | - | 2 - 3 | 180 | 92 | ||
| AK290281.1 | - | 3 - 4 | 290 | 92 | ||
| BC125158.1 | - | 2 - 3 | 151 | 40120955 | 92 | |
| BG823061.1 | - | 3 - 4 | 300 | 92 | ||
| CR541838.1 | - | 1 - 2 | 89 | 92 | ||
| Z11502.1 | - | 2 - 3 | 168 | 92 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001593.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000519045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000519045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000519045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020852.hg.1 | TC08001593.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020852.hg.1 | ENST00000262219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020852.hg.1 | ENST00000419625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020852.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020852.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179385.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179385.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179385.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179385.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179385.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179385.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179385.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179385.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179385.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179385.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179385.hg.1 | TC0800011691.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000519045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000519045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000519045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |

ANXA13
Genomic Map