Assay Details
| Gene Symbol: ANXA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003954.2 | NP_001003954.1 | 8 - 9 | 754 | 73 | |
| NM_004306.3 | NP_004297.2 | 7 - 8 | 631 | 73 | ||
| GenBank mRNA | AJ306450.1 | - | 8 - 9 | 730 | 73 | |
| AK290281.1 | - | 8 - 9 | 740 | 73 | ||
| BC125158.1 | - | 7 - 8 | 601 | 40120955 | 73 | |
| CR541838.1 | - | 6 - 7 | 539 | 73 | ||
| Z11502.1 | - | 7 - 8 | 618 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001593.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08020846.hg.1 | TC08001593.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020846.hg.1 | ENST00000262219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020846.hg.1 | ENST00000419625.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020846.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020846.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000519045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000519045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000519045.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000520591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000519045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000519045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000519045.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520519.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000520591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | ENST00000523822.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011691.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179379.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179379.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179379.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179379.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179379.hg.1 | ENST00000262219.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179379.hg.1 | ENST00000419625.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179379.hg.1 | NM_001003954.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179379.hg.1 | NM_004306.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179379.hg.1 | TC0800011691.hg.1 | Clariom D TC |

ANXA13
Genomic Map