Assay Details
| Gene Symbol: SLK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304743.1 | NP_001291672.1 | 14 - 15 | 3598 | 98 | |
| NM_014720.3 | NP_055535.2 | 15 - 16 | 3691 | 98 | ||
| XM_011540401.2 | XP_011538703.1 | 14 - 15 | 2347 | 98 | ||
| GenBank mRNA | AB002804.1 | - | 14 - 15 | 3269 | 98 | |
| AF273048.1 | - | 15 - 16 | 3164 | 98 | ||
| AK302855.1 | - | 14 - 15 | 3031 | 98 | ||
| BC111565.1 | - | 14 - 15 | 3068 | 40009070 | 98 | |
| D86959.1 | - | 14 - 15 | 3548 | 98 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000779.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
signal transduction by protein phosphorylation
regulation of cell migration
cytoplasmic microtubule organization
regulation of apoptotic process
protein autophosphorylation
regulation of focal adhesion assembly
cell-cell adhesion
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
identical protein binding
protein homodimerization activity
cadherin binding involved in cell-cell adhesion
apoptotic process
signal transduction by protein phosphorylation
regulation of cell migration
cytoplasmic microtubule organization
regulation of apoptotic process
protein autophosphorylation
regulation of focal adhesion assembly
cell-cell adhesion
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
identical protein binding
protein homodimerization activity
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009296.hg.1 | ENST00000335753.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009296.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009296.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009296.hg.1 | TC10000779.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000160186.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160186.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160186.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160186.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160186.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160186.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160186.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160186.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160186.hg.1 | TC1000008795.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |

SLK
Genomic Map