Assay Details
| Gene Symbol: SLK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304743.1 | NP_001291672.1 | 16 - 17 | 3917 | 63 | |
| NM_014720.3 | NP_055535.2 | 17 - 18 | 4010 | 63 | ||
| XM_011540401.2 | XP_011538703.1 | 16 - 17 | 2666 | 63 | ||
| GenBank mRNA | AB002804.1 | - | 16 - 17 | 3588 | 63 | |
| AF273048.1 | - | 17 - 18 | 3483 | 63 | ||
| BC111565.1 | - | 16 - 17 | 3387 | 40009070 | 63 | |
| D86959.1 | - | 16 - 17 | 3867 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
signal transduction by protein phosphorylation
regulation of cell migration
cytoplasmic microtubule organization
regulation of apoptotic process
protein autophosphorylation
regulation of focal adhesion assembly
cell-cell adhesion
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
identical protein binding
protein homodimerization activity
cadherin binding involved in cell-cell adhesion
apoptotic process
signal transduction by protein phosphorylation
regulation of cell migration
cytoplasmic microtubule organization
regulation of apoptotic process
protein autophosphorylation
regulation of focal adhesion assembly
cell-cell adhesion
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
identical protein binding
protein homodimerization activity
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | ENST00000474260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008795.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160188.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160188.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160188.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160188.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160188.hg.1 | ENST00000335753.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160188.hg.1 | ENST00000369755.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160188.hg.1 | NM_001304743.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160188.hg.1 | NM_014720.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160188.hg.1 | TC1000008795.hg.1 | Clariom D TC |

SLK
Genomic Map