Assay Details
| Gene Symbol: EIF2B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014239.3 | NP_055054.1 | 1 - 2 | 247 | 65 | |
| GenBank mRNA | AF035280.1 | - | 1 - 2 | 211 | 65 | |
| BC000494.2 | - | 1 - 2 | 213 | 2820668 | 65 | |
| BC003165.1 | - | 1 - 2 | 213 | 2820668 | 65 | |
| BC011750.2 | - | 1 - 2 | 188 | 3534548 | 65 | |
| BI908906.1 | - | 1 - 2 | 247 | 65 | ||
| BX161473.1 | - | 1 - 2 | 245 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000466.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle development
translational initiation
regulation of translational initiation
central nervous system development
response to heat
response to glucose
oligodendrocyte development
L-methionine biosynthetic process from methylthioadenosine
myelination
response to peptide hormone
positive regulation of GTPase activity
cellular response to stimulus
translation initiation factor activity
guanyl-nucleotide exchange factor activity
protein binding
ATP binding
GTP binding
S-methyl-5-thioribose-1-phosphate isomerase activity
ovarian follicle development
translational initiation
regulation of translational initiation
central nervous system development
response to heat
response to glucose
oligodendrocyte development
L-methionine biosynthetic process from methylthioadenosine
myelination
response to peptide hormone
positive regulation of GTPase activity
cellular response to stimulus
translation initiation factor activity
guanyl-nucleotide exchange factor activity
protein binding
ATP binding
GTP binding
S-methyl-5-thioribose-1-phosphate isomerase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14005830.hg.1 | ENST00000266126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005830.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005830.hg.1 | TC14000466.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005831.hg.1 | ENST00000266126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005831.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005831.hg.1 | TC14000466.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000554748.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000554748.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000554748.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000556668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000556668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000556668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000556668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | ENST00000556668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005828.hg.1 | ENST00000266126.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005828.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14005828.hg.1 | TC14000466.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400154367.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154367.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154367.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154367.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154367.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154367.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154367.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154367.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154367.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154367.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154367.hg.1 | TC1400007695.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400154368.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154368.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154368.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154368.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154368.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154368.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154368.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154368.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154368.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154368.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154368.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154368.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154368.hg.1 | TC1400007695.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400154363.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154363.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154363.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154363.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154363.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154363.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154363.hg.1 | TC1400007695.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000554748.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000554748.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000554748.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000556668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000556668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000556668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000556668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | ENST00000556668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007695.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154370.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154370.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154370.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154370.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154370.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154370.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154370.hg.1 | ENST00000266126.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154370.hg.1 | ENST00000553401.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154370.hg.1 | ENST00000553539.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154370.hg.1 | ENST00000555522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154370.hg.1 | ENST00000556028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400154370.hg.1 | NM_014239.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400154370.hg.1 | TC1400007695.hg.1 | Clariom D TC |

EIF2B2
Genomic Map