Assay Details
| Gene Symbol: PARP16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316943.1 | NP_001303872.1 | 5 - 6 | 1250 | 92 | |
| NM_001316944.1 | NP_001303873.1 | 3 - 4 | 905 | 92 | ||
| XM_006720590.2 | XP_006720653.1 | 5 - 6 | 1291 | 92 | ||
| XM_017022386.1 | XP_016877875.1 | 5 - 6 | 1234 | 92 | ||
| XM_017022387.1 | XP_016877876.1 | 4 - 5 | 1155 | 92 | ||
| XM_017022389.1 | XP_016877878.1 | 4 - 5 | 1084 | 92 | ||
| GenBank mRNA | AK000516.1 | - | 3 - 4 | 905 | 92 | |
| AK058010.1 | - | 5 - 6 | 834 | 92 | ||
More Information
Set Membership: |
|

PARP16
Genomic Map