Assay Details
| Gene Symbol: PAFAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000437.3 | NP_000428.2 | 5 - 6 | 594 | 70 | |
| XM_006710669.3 | XP_006710732.1 | 2 - 3 | 886 | 70 | ||
| XM_006710670.3 | XP_006710733.1 | 5 - 6 | 659 | 70 | ||
| XM_011541528.2 | XP_011539830.1 | 6 - 7 | 725 | 70 | ||
| XM_011541529.2 | XP_011539831.1 | 3 - 4 | 347 | 70 | ||
| XM_017001390.1 | XP_016856879.1 | 5 - 6 | 620 | 70 | ||
| XM_017001391.1 | XP_016856880.1 | 4 - 5 | 395 | 70 | ||
| XM_017001392.1 | XP_016856881.1 | 4 - 5 | 466 | 70 | ||
| GenBank mRNA | AK313815.1 | - | 5 - 6 | 521 | 70 | |
| BC001158.1 | - | 5 - 6 | 539 | 3354450 | 70 | |
| D87845.1 | - | 5 - 6 | 617 | 70 | ||
| U89386.1 | - | 5 - 6 | 493 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002384.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01036742.hg.1 | TC01002384.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01036742.hg.1 | ENST00000374282.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01036742.hg.1 | ENST00000374284.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01036742.hg.1 | ENST00000493892.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01036742.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000476611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000476611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000476611.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199318.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199318.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199318.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199318.hg.1 | ENST00000441420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199318.hg.1 | ENST00000464540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199318.hg.1 | ENST00000476611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199318.hg.1 | ENST00000493892.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199318.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199318.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199318.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199318.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199318.hg.1 | ENST00000441420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199318.hg.1 | ENST00000464540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199318.hg.1 | ENST00000476611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199318.hg.1 | ENST00000493892.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100199318.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100199318.hg.1 | TC0100013371.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374282.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000374284.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000439092.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000441420.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000464540.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000476611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000476611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000476611.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | ENST00000493892.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013371.hg.1 | NM_000437.3 | Refseq |

PAFAH2
Genomic Map