Assay Details
| Gene Symbol: ANKRD52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173595.3 | NP_775866.2 | 13 - 14 | 1459 | 72 | |
| XM_011538197.2 | XP_011536499.1 | 12 - 13 | 1561 | 72 | ||
| XM_017019183.1 | XP_016874672.1 | 12 - 13 | 1642 | 72 | ||
| XM_017019184.1 | XP_016874673.1 | 13 - 14 | 1491 | 72 | ||
| GenBank mRNA | AB190990.1 | - | 13 - 14 | 1476 | 72 | |
| AK125379.1 | - | 13 - 14 | 1476 | 72 | ||
| AK126893.1 | - | 6 - 7 | 619 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001599.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000551023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000551023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000551023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000551023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021272.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021272.hg.1 | ENST00000417002.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021272.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12021272.hg.1 | TC12001599.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200183998.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183998.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183998.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200183998.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200183998.hg.1 | TC1200010902.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000551023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000551023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000551023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000551023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |

ANKRD52
Genomic Map