Assay Details
| Gene Symbol: SPATA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286239.1 | NP_001273168.1 | 10 - 11 | 1354 | 116 | |
| NM_019073.3 | NP_061946.1 | 11 - 12 | 1396 | 116 | ||
| XM_006710699.2 | XP_006710762.1 | 11 - 12 | 1390 | 116 | ||
| XM_006710700.2 | XP_006710763.1 | 9 - 10 | 1195 | 116 | ||
| XM_011541607.2 | XP_011539909.1 | 8 - 9 | 959 | 116 | ||
| XM_011541608.1 | XP_011539910.1 | 10 - 11 | 1333 | 116 | ||
| XM_011541609.2 | XP_011539911.1 | 9 - 10 | 1041 | 116 | ||
| XM_017001510.1 | XP_016856999.1 | 8 - 9 | 1145 | 116 | ||
| XM_017001512.1 | XP_016857001.1 | 12 - 13 | 1239 | 116 | ||
| XM_017001513.1 | XP_016857002.1 | 9 - 10 | 1003 | 116 | ||
| XM_017001514.1 | XP_016857003.1 | 12 - 13 | 1281 | 116 | ||
| GenBank mRNA | AK000869.1 | - | 11 - 12 | 1361 | 116 | |
| AK301770.1 | - | 10 - 11 | 1297 | 116 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002637.hg.1
Set Membership: |
|

SPATA6
Genomic Map