Assay Details
| Gene Symbol: TOB2P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC015543.1 | - | 1 | 39 | 78 | |
| BC039027.1 | - | 1 | 57 | 78 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TOB2P1
Genomic Map