Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000492238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000492238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000492238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
ENST00000492238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035090.hg.1 |
TC02005042.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035090.hg.1 |
ENST00000376826.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035090.hg.1 |
ENST00000428343.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02035090.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200200338.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200200338.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200200338.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200200338.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200200338.hg.1 |
TC0200013669.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000428343.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000481179.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000484091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000492238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000492238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000492238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
ENST00000492238.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013669.hg.1 |
NM_001145664.1 |
Refseq |