Assay Details
| Gene Symbol: FBXL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005270151.3 | XP_005270208.1 | 1 - 2 | 186 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005270151.3 transcripts.
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
dorsal/ventral pattern formation
protein ubiquitination
bone mineralization
positive regulation of BMP signaling pathway
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
protein binding
G2/M transition of mitotic cell cycle
dorsal/ventral pattern formation
protein ubiquitination
bone mineralization
positive regulation of BMP signaling pathway
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000425536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000425536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000425536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000425536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000425536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000425536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000432590.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000432590.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000432590.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000432590.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000432590.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000432590.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000440407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000440407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000440407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000440407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000440407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000440407.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000457067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000457067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000457067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000457067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000457067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000457067.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000481808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000481808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000481808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | ENST00000481808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000224862.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000369956.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000425536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000425536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000425536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000425536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000425536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000425536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000432590.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000432590.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000432590.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000432590.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000432590.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000432590.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000440407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000440407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000440407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000440407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000440407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000440407.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000457067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000457067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000457067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000457067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000457067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000457067.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000481808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000481808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000481808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | ENST00000481808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | NM_024326.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008728.hg.1 | NM_024326.3 | Refseq |

FBXL15
Genomic Map