Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001447.hg.1 |
ENST00000361728.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001447.hg.1 |
ENST00000389232.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001447.hg.1 |
ENST00000415757.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001447.hg.1 |
NM_001036.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001447.hg.1 |
NM_001243996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15001447.hg.1 |
TC15000229.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500147351.hg.1 |
ENST00000389232.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147351.hg.1 |
ENST00000415757.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147351.hg.1 |
ENST00000622037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147351.hg.1 |
NM_001036.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147351.hg.1 |
NM_001243996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147351.hg.1 |
ENST00000389232.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147351.hg.1 |
ENST00000415757.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147351.hg.1 |
ENST00000622037.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500147351.hg.1 |
NM_001036.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147351.hg.1 |
NM_001243996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500147351.hg.1 |
TC1500006806.hg.1 |
Clariom D TC |