Assay Details
| Gene Symbol: NIFK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032390.4 | NP_115766.3 | 2 - 3 | 327 | 63 | |
| GenBank mRNA | AB044971.1 | - | 2 - 3 | 304 | 63 | |
| AK291903.1 | - | 2 - 3 | 295 | 63 | ||
| AK299814.1 | - | 2 - 3 | 277 | 63 | ||
| AK311033.1 | - | 2 - 3 | 272 | 63 | ||
| BC012457.1 | - | 1 - 2 | 52 | 63 | ||
| BC022990.1 | - | 2 - 3 | 283 | 4997441 | 63 | |
| BC024238.2 | - | 2 - 3 | 292 | 4797633 | 63 | |
| DA570244.1 | - | 2 - 3 | 327 | 63 | ||
More Information
Set Membership: |
|

NIFK
Genomic Map