Assay Details
| Gene Symbol: NIFK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032390.4 | NP_115766.3 | 4 - 5 | 632 | 147 | |
| GenBank mRNA | AB044971.1 | - | 4 - 5 | 609 | 147 | |
| AK291903.1 | - | 4 - 5 | 600 | 147 | ||
| BC012457.1 | - | 3 - 4 | 357 | 147 | ||
| BC022990.1 | - | 4 - 5 | 588 | 4997441 | 147 | |
| BC024238.2 | - | 4 - 5 | 597 | 4797633 | 147 | |
| DA570244.1 | - | 4 - 5 | 632 | 147 | ||
More Information
Set Membership: |
|

NIFK
Genomic Map