Assay Details
| Gene Symbol: TOMM70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014820.4 | NP_055635.3 | 1 - 2 | 774 | 65 | |
| GenBank mRNA | AB018262.1 | - | 1 - 2 | 416 | 65 | |
| BC003633.2 | - | 1 - 2 | 440 | 3605218 | 65 | |
| BC052994.1 | - | 1 - 2 | 757 | 5549394 | 65 | |
| BC065555.1 | - | 1 - 2 | 771 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001611.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03029049.hg.1 | TC03001611.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029049.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029049.hg.1 | ENST00000544924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029049.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000483945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000483945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000483945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000483945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000492171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000492171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | ENST00000492171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029048.hg.1 | TC03001611.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029048.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029048.hg.1 | ENST00000544924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029048.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029051.hg.1 | TC03001611.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029051.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029051.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029050.hg.1 | TC03001611.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029050.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029050.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029052.hg.1 | TC03001611.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029052.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029052.hg.1 | ENST00000544924.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03029052.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187969.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187969.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187969.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187969.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187969.hg.1 | TC0300011842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300187970.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187970.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187970.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187970.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187970.hg.1 | TC0300011842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300187972.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187972.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187972.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187972.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187972.hg.1 | TC0300011842.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000483945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000483945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000483945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000483945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000492171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000492171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | ENST00000492171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011842.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187971.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187971.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187971.hg.1 | ENST00000284320.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187971.hg.1 | NM_014820.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187971.hg.1 | TC0300011842.hg.1 | Clariom D TC |

TOMM70
Genomic Map