| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029046.hg.1 |
TC03001611.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029046.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029046.hg.1 |
ENST00000544924.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03029046.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000483945.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000483945.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000483945.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000483945.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000492171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000492171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
ENST00000492171.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300187967.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300187967.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300187967.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300187967.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300187967.hg.1 |
TC0300011842.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000284320.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000483945.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000483945.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000483945.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000483945.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000492171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000492171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
ENST00000492171.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011842.hg.1 |
NM_014820.4 |
Refseq |