Assay Details
| Gene Symbol: KLRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016523.2 | NP_057607.1 | 2 - 3 | 253 | 115 | |
| XM_017019414.1 | XP_016874903.1 | 2 - 3 | 372 | 115 | ||
| GenBank mRNA | AF175206.1 | - | 2 - 3 | 253 | 115 | |
| AJ305370.1 | - | 2 - 3 | 236 | 115 | ||
| BC096734.1 | - | 2 - 3 | 213 | 115 | ||
| BC098166.1 | - | 2 - 3 | 213 | 40015535 | 115 | |
| BC098257.1 | - | 2 - 3 | 213 | 40015534 | 115 | |
| BC098354.1 | - | 2 - 3 | 213 | 40015531 | 115 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000152.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NR_120305.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NR_120305.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NR_120305.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NR_120305.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NR_120305.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NR_120305.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006773.hg.1 | NR_120305.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001965.hg.1 | ENST00000279544.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001965.hg.1 | ENST00000537723.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001965.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12001965.hg.1 | TC12000152.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200147599.hg.1 | ENST00000537723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147599.hg.1 | ENST00000545196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147599.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147599.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147599.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200147599.hg.1 | ENST00000537723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147599.hg.1 | ENST00000545196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147599.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147599.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147599.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200147599.hg.1 | TC1200006773.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000279545.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000354855.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000537723.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000545196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000612321.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000616259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617793.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | ENST00000617889.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291822.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_001291823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NM_016523.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NR_120305.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NR_120305.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NR_120305.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NR_120305.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NR_120305.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NR_120305.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006773.hg.1 | NR_120305.1 | Refseq |

KLRF1
Genomic Map