Assay Details
| Gene Symbol: ELP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288725.1 | NP_001275654.1 | 3 - 4 | 436 | 155 | |
| NM_001288726.1 | NP_001275655.1 | 3 - 4 | 436 | 155 | ||
| NM_019040.4 | NP_061913.3 | 3 - 4 | 436 | 155 | ||
| GenBank mRNA | AK000505.1 | - | 3 - 4 | 415 | 155 | |
| AK304885.1 | - | 3 - 4 | 395 | 155 | ||
| AL136677.1 | - | 3 - 4 | 393 | 155 | ||
| BC012514.1 | - | 3 - 4 | 387 | 4509903 | 155 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000303.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
histone H3 acetylation
histone H4 acetylation
regulation of protein kinase activity
RNA polymerase II core binding
histone acetyltransferase activity
protein binding
phosphorylase kinase regulator activity
regulation of transcription from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
histone H3 acetylation
histone H4 acetylation
regulation of protein kinase activity
RNA polymerase II core binding
histone acetyltransferase activity
protein binding
phosphorylase kinase regulator activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003859.hg.1 | ENST00000350638.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003859.hg.1 | ENST00000379163.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003859.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003859.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003859.hg.1 | TC11000303.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100150536.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150536.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150536.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150536.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150536.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150536.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150536.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150536.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150536.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150536.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150536.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100150536.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150536.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150536.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100150536.hg.1 | TC1100007185.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000350638.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000379163.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000395934.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | ENST00000474374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288725.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_001288726.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_019040.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007185.hg.1 | NM_019040.4 | Refseq |

ELP4
Genomic Map