Assay Details
| Gene Symbol: CNTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK023327.1 | - | 1 - 2 | 284 | 88 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000221818.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000221818.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000221818.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000221818.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000221818.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000221818.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000221818.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000221818.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000513948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000513948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000513948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000513948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000513948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000513948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000593335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000593335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000593335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000599263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000599263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | ENST00000599263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000221818.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000221818.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000221818.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000221818.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000221818.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000221818.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000221818.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000221818.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000430325.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000513948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000513948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000513948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000513948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000513948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000513948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000593335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000593335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000593335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000599263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000599263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | ENST00000599263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010695.hg.1 | NM_024877.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 208992 | - | - |

CNTD2
Genomic Map