Assay Details
| Gene Symbol: CHRM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000741.3 | NP_000732.2 | 1 | 1596 | 70 | |
| GenBank mRNA | BF724332.1 | - | 1 | 199 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

CHRM4
Genomic Map