Assay Details
Gene Symbol: PLA2G4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206670.1 | NP_001193599.1 | 14 - 15 | 1581 | 94 | |
XM_011521237.2 | XP_011519539.1 | 14 - 15 | 1491 | 94 | ||
XM_011521238.1 | XP_011519540.1 | 5 - 6 | 1299 | 94 | ||
XM_011521239.1 | XP_011519541.1 | 3 - 4 | 427 | 94 | ||
GenBank mRNA | AK127558.1 | - | 4 - 5 | 870 | 94 | |
BC101584.1 | - | 3 - 4 | 486 | 94 | ||
BC101612.1 | - | 3 - 4 | 486 | 94 | ||
BX648482.1 | - | 4 - 5 | 780 | 94 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15001258.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phospholipid metabolic process
phospholipid catabolic process
phosphatidylinositol acyl-chain remodeling
phosphatidylserine acyl-chain remodeling
phosphatidylcholine acyl-chain remodeling
phosphatidylethanolamine acyl-chain remodeling
lysophospholipase activity
phospholipase A2 activity
phosphatidylcholine 1-acylhydrolase activity
metal ion binding
phospholipid metabolic process
phospholipid catabolic process
phosphatidylinositol acyl-chain remodeling
phosphatidylserine acyl-chain remodeling
phosphatidylcholine acyl-chain remodeling
phosphatidylethanolamine acyl-chain remodeling
lysophospholipase activity
phospholipase A2 activity
phosphatidylcholine 1-acylhydrolase activity
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR15011492.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15011492.hg.1 | ENST00000413860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15011492.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15011492.hg.1 | TC15001258.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15011494.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15011494.hg.1 | ENST00000413860.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15011494.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15011494.hg.1 | TC15001258.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | ENST00000551073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009177.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500166539.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500166539.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500166539.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500166539.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500166539.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500166539.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500166539.hg.1 | TC1500009177.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500166537.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500166537.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500166537.hg.1 | ENST00000551073.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500166537.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500166537.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500166537.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500166537.hg.1 | ENST00000551073.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500166537.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500166537.hg.1 | TC1500009177.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500166538.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500166538.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500166538.hg.1 | ENST00000551073.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500166538.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500166538.hg.1 | ENST00000399518.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500166538.hg.1 | ENST00000547930.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500166538.hg.1 | ENST00000551073.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500166538.hg.1 | NM_001206670.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500166538.hg.1 | TC1500009177.hg.1 | Clariom D TC |
