Assay Details
| Gene Symbol: ALPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020778.4 | NP_065829.3 | 1 - 2 | 917 | 69 | |
| XM_017022445.1 | XP_016877934.1 | 1 - 2 | 966 | 69 | ||
| GenBank mRNA | AY044449.1 | - | 1 - 2 | 917 | 69 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000822.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15007485.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15007485.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15007485.hg.1 | TC15000822.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000558077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000558077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000558077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | ENST00000558077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15007484.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15007484.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15007484.hg.1 | TC15000822.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000558077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000558077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000558077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | ENST00000558077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008170.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500158789.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500158789.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500158789.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500158789.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500158789.hg.1 | TC1500008170.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500158788.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500158788.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500158788.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500158788.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500158788.hg.1 | TC1500008170.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500158787.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500158787.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500158787.hg.1 | ENST00000258888.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500158787.hg.1 | NM_020778.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500158787.hg.1 | TC1500008170.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 195121 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 205382 | - | - |

ALPK3
Genomic Map