Assay Details
| Gene Symbol: HDAC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098416.3 | NP_001091886.1 | 8 - 9 | 987 | 49 | |
| XM_011538480.1 | XP_011536782.1 | 8 - 9 | 939 | 49 | ||
| GenBank mRNA | AK024469.1 | - | 4 - 5 | 413 | 49 | |
| AY321367.1 | - | 8 - 9 | 872 | 49 | ||
| BC064840.1 | - | 8 - 9 | 881 | 6179239 | 49 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001438.hg.1
Set Membership: |
|

HDAC7
Genomic Map