Assay Details
| Gene Symbol: NGLY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145294.1 | NP_001138766.1 | 7 - 8 | 1198 | 77 | |
| NM_001145295.1 | NP_001138767.1 | 7 - 8 | 1258 | 77 | ||
| NM_018297.3 | NP_060767.2 | 7 - 8 | 1258 | 77 | ||
| XM_005265316.1 | XP_005265373.1 | 7 - 8 | 1203 | 77 | ||
| XM_005265317.1 | XP_005265374.1 | 7 - 8 | 1203 | 77 | ||
| XM_011533944.1 | XP_011532246.1 | 7 - 8 | 1154 | 77 | ||
| XM_017006839.1 | XP_016862328.1 | 7 - 8 | 1203 | 77 | ||
| GenBank mRNA | AF250924.2 | - | 7 - 8 | 1191 | 77 | |
| AK001867.1 | - | 5 - 6 | 859 | 77 | ||
| AK296047.1 | - | 7 - 8 | 1165 | 77 | ||
| BC007226.1 | - | 7 - 8 | 1191 | 2963554 | 77 | |
| BC017220.2 | - | 7 - 8 | 1185 | 4301811 | 77 | |
More Information
Set Membership: |
|

NGLY1
Genomic Map