Assay Details
| Gene Symbol: C14orf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127393.1 | NP_001120865.1 | 2 - 3 | 364 | 92 | |
| GenBank mRNA | BX161388.1 | - | 2 - 3 | 299 | 92 | |
| CB127906.1 | - | 2 - 3 | 323 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000553449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000553449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554713.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554713.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554713.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400176428.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176428.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400176428.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176428.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400176428.hg.1 | TC1400010339.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000553449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000553449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554713.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554713.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554713.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_004894.2 | Refseq |

C14orf2
Genomic Map