Assay Details
| Gene Symbol: MYH14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006723386.3 | XP_006723449.1 | 1 - 2 | 110 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006723386.3 transcripts.
Set Membership: |
|

MYH14
Genomic Map