Assay Details
| Gene Symbol: HCRTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017001105.1 | XP_016856594.1 | 2 - 3 | 213 | 78 | |
| XM_017001106.1 | XP_016856595.1 | 1 - 2 | 103 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017001105.1,XM_017001106.1 transcripts.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
neuropeptide signaling pathway
chemical synaptic transmission
feeding behavior
cellular response to hormone stimulus
regulation of circadian sleep/wake cycle, sleep
regulation of cytosolic calcium ion concentration
response to peptide
G-protein coupled receptor activity
neuropeptide receptor activity
orexin receptor activity
peptide hormone binding
peptide binding
neuropeptide signaling pathway
chemical synaptic transmission
feeding behavior
cellular response to hormone stimulus
regulation of circadian sleep/wake cycle, sleep
regulation of cytosolic calcium ion concentration
response to peptide
G-protein coupled receptor activity
neuropeptide receptor activity
orexin receptor activity
peptide hormone binding
peptide binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |

HCRTR1
Genomic Map