Assay Details
| Gene Symbol: HCRTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001525.2 | NP_001516.2 | 6 - 7 | 1125 | 100 | |
| XM_017001105.1 | XP_016856594.1 | 6 - 7 | 1393 | 100 | ||
| XM_017001106.1 | XP_016856595.1 | 6 - 7 | 1049 | 100 | ||
| XM_017001107.1 | XP_016856596.1 | 4 - 5 | 1616 | 100 | ||
| GenBank mRNA | AF041243.1 | - | 4 - 5 | 893 | 100 | |
| AK290521.1 | - | 6 - 7 | 1125 | 100 | ||
| BC074796.2 | - | 4 - 5 | 782 | 30915259 | 100 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000412.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
neuropeptide signaling pathway
chemical synaptic transmission
feeding behavior
cellular response to hormone stimulus
regulation of circadian sleep/wake cycle, sleep
regulation of cytosolic calcium ion concentration
response to peptide
G-protein coupled receptor activity
neuropeptide receptor activity
orexin receptor activity
peptide hormone binding
peptide binding
neuropeptide signaling pathway
chemical synaptic transmission
feeding behavior
cellular response to hormone stimulus
regulation of circadian sleep/wake cycle, sleep
regulation of cytosolic calcium ion concentration
response to peptide
G-protein coupled receptor activity
neuropeptide receptor activity
orexin receptor activity
peptide hormone binding
peptide binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006306.hg.1 | TC01000412.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006306.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006306.hg.1 | ENST00000373706.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006306.hg.1 | ENST00000403528.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006306.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006306.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006307.hg.1 | TC01000412.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006307.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006307.hg.1 | ENST00000373706.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006307.hg.1 | ENST00000403528.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006307.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01006307.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153638.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153638.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153638.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153638.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153638.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153638.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153638.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153638.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153638.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153638.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153638.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153638.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153638.hg.1 | TC0100007646.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007646.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153639.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153639.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153639.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153639.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153639.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153639.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153639.hg.1 | ENST00000373705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153639.hg.1 | ENST00000373706.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153639.hg.1 | ENST00000403528.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153639.hg.1 | ENST00000468521.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153639.hg.1 | ENST00000485464.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100153639.hg.1 | NM_001525.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100153639.hg.1 | TC0100007646.hg.1 | Clariom D TC |

HCRTR1
Genomic Map