Assay Details
| Gene Symbol: TIA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022173.2 | NP_071505.2 | 3 - 4 | 434 | 145 | |
| XM_005264528.4 | XP_005264585.1 | 2 - 3 | 346 | 145 | ||
| XM_005264531.1 | XP_005264588.1 | 3 - 4 | 362 | 145 | ||
| XM_011533077.1 | XP_011531379.1 | 3 - 4 | 444 | 145 | ||
| XM_011533078.1 | XP_011531380.1 | 3 - 4 | 362 | 145 | ||
| XM_011533081.2 | XP_011531383.1 | 3 - 4 | 443 | 145 | ||
| XM_017004789.1 | XP_016860278.1 | 3 - 4 | 263 | 145 | ||
| XM_017004792.1 | XP_016860281.1 | 3 - 4 | 443 | 145 | ||
| XM_017004793.1 | XP_016860282.1 | 3 - 4 | 443 | 145 | ||
| XM_017004795.1 | XP_016860284.1 | 3 - 4 | 322 | 145 | ||
| XM_017004796.1 | XP_016860285.1 | 3 - 4 | 443 | 145 | ||
| XM_017004797.1 | XP_016860286.1 | 3 - 4 | 443 | 145 | ||
| GenBank mRNA | AB209211.1 | - | 3 - 4 | 445 | 145 | |
| BC015944.1 | - | 3 - 4 | 241 | 3909177 | 145 | |
| CX867570.1 | - | 3 - 4 | 407 | 145 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the XM_005264528.4,XM_005264531.1,XM_011533078.1,XM_011533081.2,XM_017004792.1,XM_017004793.1,XM_017004795.1,XM_017004796.1,XM_017004797.1 transcripts.
This assay is in the 5'UTR region of the XM_005264528.4,XM_005264531.1,XM_011533078.1,XM_011533081.2,XM_017004792.1,XM_017004793.1,XM_017004795.1,XM_017004796.1,XM_017004797.1 transcripts.
Set Membership: |
|

TIA1
Genomic Map