Assay Details
| Gene Symbol: OPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015560.2 | NP_056375.2 | 5 - 6 | 864 | 78 | |
| NM_130831.2 | NP_570844.1 | 4 - 5 | 756 | 78 | ||
| NM_130832.2 | NP_570845.1 | 5 - 6 | 810 | 78 | ||
| NM_130834.2 | NP_570847.2 | 6 - 7 | 918 | 78 | ||
| GenBank mRNA | AB011139.1 | - | 5 - 6 | 685 | 78 | |
| BC075805.1 | - | 5 - 6 | 784 | 4214698 | 78 | |
More Information
Set Membership: |
|

OPA1
Genomic Map