Assay Details
| Gene Symbol: NR2F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145155.1 | NP_001138627.1 | 1 - 2 | 430 | 65 | |
| GenBank mRNA | AK301595.1 | - | 1 - 2 | 67 | 65 | |
| BC106083.1 | - | 1 - 2 | 430 | 4453629 | 65 | |
| FM208183.1 | - | 1 - 2 | 420 | 65 | ||
| HQ692849.1 | - | 1 - 2 | 48 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000936.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
neuron migration
maternal placenta development
negative regulation of endothelial cell proliferation
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
lipid metabolic process
signal transduction
skeletal muscle tissue development
fertilization
anterior/posterior pattern specification
radial pattern formation
negative regulation of endothelial cell migration
intracellular receptor signaling pathway
forebrain development
response to estradiol
steroid hormone mediated signaling pathway
negative regulation of cyclin-dependent protein serine/threonine kinase activity
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
blood vessel morphogenesis
limb development
placenta blood vessel development
trophoblast giant cell differentiation
regulation of transcription involved in lymphatic endothelial cell fate commitment
retinoic acid binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
transcription corepressor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
protein binding
zinc ion binding
protein homodimerization activity
sequence-specific DNA binding
negative regulation of transcription from RNA polymerase II promoter
neuron migration
maternal placenta development
negative regulation of endothelial cell proliferation
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
lipid metabolic process
signal transduction
skeletal muscle tissue development
fertilization
anterior/posterior pattern specification
radial pattern formation
negative regulation of endothelial cell migration
intracellular receptor signaling pathway
forebrain development
response to estradiol
steroid hormone mediated signaling pathway
negative regulation of cyclin-dependent protein serine/threonine kinase activity
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
blood vessel morphogenesis
limb development
placenta blood vessel development
trophoblast giant cell differentiation
regulation of transcription involved in lymphatic endothelial cell fate commitment
retinoic acid binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
transcription corepressor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
protein binding
zinc ion binding
protein homodimerization activity
sequence-specific DNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15008718.hg.1 | ENST00000421109.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008718.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008718.hg.1 | TC15000936.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000453270.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000453270.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000453270.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000453270.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000453270.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000453270.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000453270.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000559679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | ENST00000559679.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500161294.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500161294.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500161294.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500161294.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500161294.hg.1 | TC1500008442.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394166.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000394171.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000453270.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000453270.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000453270.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000453270.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000453270.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000453270.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000453270.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000559679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | ENST00000559679.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145156.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_001145157.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008442.hg.1 | NM_021005.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500161293.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500161293.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500161293.hg.1 | ENST00000421109.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500161293.hg.1 | NM_001145155.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500161293.hg.1 | TC1500008442.hg.1 | Clariom D TC |

NR2F2
Genomic Map