Assay Details
| Gene Symbol: RDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152443.2 | NP_689656.2 | 2 - 3 | 103 | 100 | |
| GenBank mRNA | AK315462.1 | - | 2 - 3 | 103 | 100 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000410.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK315462.1,NM_152443.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14004840.hg.1 | ENST00000551171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004840.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004840.hg.1 | TC14000410.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000547463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000547463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000547463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000552873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000552873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | ENST00000552873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004848.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004848.hg.1 | ENST00000539142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004848.hg.1 | ENST00000551171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004848.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004848.hg.1 | TC14000410.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004847.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004847.hg.1 | ENST00000551171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004847.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004847.hg.1 | TC14000410.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004845.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004845.hg.1 | ENST00000547463.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004845.hg.1 | ENST00000551171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004845.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14004845.hg.1 | TC14000410.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000547463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000547463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000547463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000552873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000552873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | ENST00000552873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007504.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152647.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152647.hg.1 | ENST00000547463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152647.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152647.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152647.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152647.hg.1 | ENST00000547463.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152647.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152647.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152647.hg.1 | TC1400007504.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400152649.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152649.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152649.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152649.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152649.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152649.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152649.hg.1 | TC1400007504.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400152650.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152650.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152650.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152650.hg.1 | ENST00000267502.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152650.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152650.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152650.hg.1 | TC1400007504.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400152640.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152640.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152640.hg.1 | ENST00000551171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400152640.hg.1 | NM_152443.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400152640.hg.1 | TC1400007504.hg.1 | Clariom D TC |

RDH12
Genomic Map