Assay Details
| Gene Symbol: LRP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK291030.1 | - | 4 - 5 | 550 | 88 | |
| AL834518.1 | - | 4 - 5 | 555 | 88 | ||
| AY358399.1 | - | 4 - 5 | 950 | 88 | ||
| BC064901.1 | - | 1 - 2 | 109 | 88 | ||
| BC113714.1 | - | 4 - 5 | 432 | 8322766 | 88 | |
| BC113716.1 | - | 4 - 5 | 432 | 8322768 | 88 | |
| BX161500.1 | - | 4 - 5 | 878 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145553.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145553.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145553.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145553.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145553.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145553.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145553.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145553.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145553.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400145553.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400145553.hg.1 | TC1400006663.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000359591.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000470660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000546834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000551466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | ENST00000553002.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400006663.hg.1 | NM_014045.4 | Refseq |

LRP10
Genomic Map