| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17017860.hg.1 |
ENST00000302362.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17017860.hg.1 |
ENST00000378634.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17017860.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17017860.hg.1 |
NM_018405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17017860.hg.1 |
TC17001345.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000579741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000579741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000579741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000579741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000579984.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000579984.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
ENST00000579984.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
NM_018405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
NM_018405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
NM_018405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
NM_018405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
NM_018405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
NM_018405.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010327.hg.1 |
NM_018405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000579741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000579741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000579741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000579741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000579984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000579984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
ENST00000579984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
NM_018405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
NM_018405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
NM_018405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
NM_018405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
NM_018405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
NM_018405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010327.hg.1 |
NM_018405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700181125.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700181125.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700181125.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700181125.hg.1 |
ENST00000579741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700181125.hg.1 |
ENST00000579984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700181125.hg.1 |
NM_018405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700181125.hg.1 |
ENST00000302362.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700181125.hg.1 |
ENST00000378634.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700181125.hg.1 |
ENST00000496655.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700181125.hg.1 |
ENST00000579741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700181125.hg.1 |
ENST00000579984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700181125.hg.1 |
NM_018405.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700181125.hg.1 |
TC1700010327.hg.1 |
Clariom D TC |