Assay Details
| Gene Symbol: CNPY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318847.1 | NP_001305776.1 | 3 - 4 | 777 | 69 | |
| NR_134880.1 | 3 - 4 | 777 | 69 | |||
| NR_134881.1 | 4 - 5 | 1970 | 69 | |||
| NR_134882.1 | 2 - 3 | 653 | 69 | |||
| GenBank mRNA | AK124196.1 | - | 4 - 5 | 1969 | 69 | |
| BG675987.1 | - | 3 - 4 | 371 | 69 | ||
| BI909527.1 | - | 2 - 3 | 305 | 69 | ||
| CK429647.1 | - | 3 - 4 | 390 | 69 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

CNPY3
Genomic Map