Assay Details
| Gene Symbol: GABARAPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK311266.1 | - | 2 - 3 | 185 | 103 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000037243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000037243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000037243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000037243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000037243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000037243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000037243.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000563744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000563744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000563744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000563744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000563744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000563744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000565057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000565057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000565057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000565057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000565057.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000565985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000565985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000565985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000565985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000568455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000568455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000568455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000568455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | ENST00000568455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | NM_007285.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | NM_007285.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | NM_007285.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | NM_007285.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | NM_007285.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | NM_007285.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008454.hg.1 | NM_007285.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000037243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000037243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000037243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000037243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000037243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000037243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000037243.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000563744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000563744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000563744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000563744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000563744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000563744.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000565057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000565057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000565057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000565057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000565057.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000565985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000565985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000565985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000565985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000568455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000568455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000568455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000568455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | ENST00000568455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | NM_007285.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | NM_007285.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | NM_007285.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | NM_007285.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | NM_007285.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | NM_007285.6 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008454.hg.1 | NM_007285.6 | Refseq |

GABARAPL2
Genomic Map