Assay Details
| Gene Symbol: FAM204A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005270024.1 | XP_005270081.1 | 3 - 4 | 439 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005270024.1 transcripts.
Set Membership: |
|

FAM204A
Genomic Map