Assay Details
| Gene Symbol: GRID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017551.2 | NP_060021.1 | 15 - 16 | 2703 | 76 | |
| GenBank mRNA | AB033046.1 | - | 12 - 13 | 1950 | 76 | |
| AK127168.1 | - | 15 - 16 | 2688 | 76 | ||
| AK293104.1 | - | 15 - 16 | 2661 | 76 | ||
| AK302192.1 | - | 8 - 9 | 1475 | 76 | ||
| BC039263.1 | - | 14 - 15 | 2536 | 5273869 | 76 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001475.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10018666.hg.1 | ENST00000327946.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018666.hg.1 | ENST00000536331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018666.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018666.hg.1 | TC10001475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018668.hg.1 | ENST00000327946.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018668.hg.1 | ENST00000536331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018668.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018668.hg.1 | TC10001475.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000552278.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000552278.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000552278.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000614988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000614988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | ENST00000614988.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018664.hg.1 | ENST00000327946.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018664.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10018664.hg.1 | TC10001475.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000176769.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176769.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176769.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176769.hg.1 | ENST00000552278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176769.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176769.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176769.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176769.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176769.hg.1 | ENST00000552278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176769.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176769.hg.1 | TC1000011287.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000552278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000552278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000552278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000614988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000614988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | ENST00000614988.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011287.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176767.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176767.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176767.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176767.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176767.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176767.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176767.hg.1 | TC1000011287.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000176770.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176770.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176770.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176770.hg.1 | ENST00000552278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176770.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176770.hg.1 | ENST00000327946.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176770.hg.1 | ENST00000464741.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176770.hg.1 | ENST00000536331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176770.hg.1 | ENST00000552278.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000176770.hg.1 | NM_017551.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000176770.hg.1 | TC1000011287.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 192681 | - | - |

GRID1
Genomic Map