Assay Details
| Gene Symbol: BACE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138992.2 | NP_620477.1 | 7 - 8 | 1598 | 70 | |
| GenBank mRNA | AF188277.1 | - | 7 - 8 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000185.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000328735.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000328735.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000328735.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000328735.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000328735.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000328735.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000328735.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000328735.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000328735.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000328735.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000328735.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000328735.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000328735.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000328735.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000328735.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000328735.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000330333.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000330333.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000330333.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000330333.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000330333.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000330333.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000330333.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000330333.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000330333.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000330333.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000330333.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000330333.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000330333.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000330333.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000330333.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000330333.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000330333.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000347667.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000463674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000463674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000463674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000463674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000463674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000463674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000463674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000463674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000463674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000463674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000465326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000465326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000465326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000465326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000465326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000465326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000465326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000465326.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000466122.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000466122.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000466122.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000466122.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000466122.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000466122.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000466122.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000466122.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000466122.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000466122.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000466122.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000466122.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000466122.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000470864.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000470864.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000470864.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000470864.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000475618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000475618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000487994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000487994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000487994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000487994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000487994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000487994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000487994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000487994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000487994.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000491838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000491838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000491838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000491838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000491838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000491838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | ENST00000491838.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_012105.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138991.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007198.hg.1 | NM_138992.2 | Refseq |

BACE2
Genomic Map