Assay Details
| Gene Symbol: DMKN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033746.1 | 13 - 14 | 719 | 64 | ||
| XM_011527495.2 | XP_011525797.1 | 22 - 23 | 1852 | 64 | ||
| XM_011527498.2 | XP_011525800.1 | 21 - 22 | 1810 | 64 | ||
| GenBank mRNA | BC011886.2 | - | 13 - 14 | 648 | 64 | |
More Information
Set Membership: |
|

DMKN
Genomic Map